ENA428682533 Enamine

2-(4-methyl-4H-1,2,4-triazol-3-yl)morpholine dihydrochloride

  • MDL number MFCD28118394
  • Cas Number : 1795502-55-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-182935

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-182935
Availability details for
Quantity
  • Availability
  • Add to Cart