ENA428682539 Enamine

5-(1,3-dioxolan-2-yl)-1,3-thiazole-2-carbaldehyde

  • MDL number MFCD28118400
  • Cas Number : 1803203-53-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-183071

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-183071
Availability details for
Quantity
  • Availability
  • Add to Cart