ENA428682540 Enamine

[4-(ethanesulfonyl)phenyl]methanamine hydrochloride

  • MDL number MFCD28118402
  • Cas Number : 98959-89-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-183083

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-183083
Availability details for
Quantity
  • Availability
  • Add to Cart