ENA428682544 Enamine

N-[2-fluoro-5-(trifluoromethyl)phenyl]guanidine hydrochloride

  • MDL number MFCD28118407
  • Cas Number : 1803587-42-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-183137

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-183137
Availability details for
Quantity
  • Availability
  • Add to Cart