ENA428682549 Enamine

ethyl 2-(propylamino)propanoate hydrochloride

  • MDL number MFCD28118412

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-183165

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-183165
Availability details for
Quantity
  • Availability
  • Add to Cart