ENA428682552 Enamine

tert-butyl 3-[2-(benzyloxy)-2-oxoethylidene]azetidine-1-carboxylate

  • MDL number MFCD28118416
  • Cas Number : 1803588-48-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-183216

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-183216
Availability details for
Quantity
  • Availability
  • Add to Cart