ENA428682553 Enamine

N-(2-aminoethyl)-N-methyloxan-4-amine dihydrochloride

  • MDL number MFCD28118419
  • Cas Number : 1803590-82-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-183234

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-183234
Availability details for
Quantity
  • Availability
  • Add to Cart