ENA428682562 Enamine

3-[4-(trifluoromethyl)phenyl]pentan-3-amine hydrochloride

  • MDL number MFCD28118432
  • Cas Number : 667916-89-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-183325

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-183325
Availability details for
Quantity
  • Availability
  • Add to Cart