ENA428682565 Enamine

2-methyl-1-(pyrrolidin-3-yl)-4,5,6,7-tetrahydro-1H-1,3-benzodiazole dihydrochloride

  • MDL number MFCD28118435
  • Cas Number : 1798745-74-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-183344

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-183344
Availability details for
Quantity
  • Availability
  • Add to Cart