ENA428682566 Enamine

2-benzyl-decahydrocyclohepta[c]pyrrol-4-one

  • MDL number MFCD28118436
  • Cas Number : 1486520-08-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-183363

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-183363
Availability details for
Quantity
  • Availability
  • Add to Cart