ENA428682570 Enamine

(1S)-2-amino-1-(4-methylphenyl)ethan-1-ol hydrochloride

  • MDL number MFCD28118447

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-183679

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-183679
Availability details for
Quantity
  • Availability
  • Add to Cart