ENA428682571 Enamine

3-(3-aminopropyl)phenol hydrochloride

  • MDL number MFCD28118448
  • Cas Number : 55260-46-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-183680

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-183680
Availability details for
Quantity
  • Availability
  • Add to Cart