ENA428682582 Enamine

2-{octahydro-1H-cyclopenta[b]pyridin-1-yl}ethan-1-amine dihydrochloride

  • MDL number MFCD28118460
  • Cas Number : 1803606-90-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-184071

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-184071
Availability details for
Quantity
  • Availability
  • Add to Cart