ENA428682584 Enamine

N-(azepan-4-yl)-N-cyclobutylmethanesulfonamide hydrochloride

  • MDL number MFCD28118468
  • Cas Number : 1803590-13-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-184084

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-184084
Availability details for
Quantity
  • Availability
  • Add to Cart