ENA428682590 Enamine

methyl 3-amino-2-(4-fluorophenyl)-2-methylpropanoate hydrochloride

  • MDL number MFCD28118476
  • Cas Number : 1795534-67-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-184278

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-184278
Availability details for
Quantity
  • Availability
  • Add to Cart