ENA428682592 Enamine

tert-butyl 4-(3-nitro-1H-pyrazol-1-yl)piperidine-1-carboxylate

  • MDL number MFCD28118478
  • Cas Number : 1201935-68-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-184299

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-184299
Availability details for
Quantity
  • Availability
  • Add to Cart