ENA428682593 Enamine

3-amino-2-hydroxypropanamide hydrochloride

  • MDL number MFCD28118479
  • Cas Number : 1795284-20-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-184355

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-184355
Availability details for
Quantity
  • Availability
  • Add to Cart