ENA428682596 Enamine

5-methyl-4,5,6,7-tetrahydro-1H-indazole-7-carboxylic acid hydrochloride

  • MDL number MFCD28118482
  • Cas Number : 1795362-25-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-184382

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-184382
Availability details for
Quantity
  • Availability
  • Add to Cart