ENA428682606 Enamine

7-bromoheptan-1-amine hydrobromide

  • MDL number MFCD28118492
  • Cas Number : 1082678-45-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-184415

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-184415
Availability details for
Quantity
  • Availability
  • Add to Cart