ENA428682609 Enamine

2-(3-amino-5-methylphenoxy)ethan-1-ol hydrochloride

  • MDL number MFCD28118496
  • Cas Number : 1795503-75-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-184469

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-184469
Availability details for
Quantity
  • Availability
  • Add to Cart