ENA428682610 Enamine

2-[3-(trifluoromethyl)-1H-pyrazol-4-yl]ethane-1-sulfonyl chloride

  • MDL number MFCD28118497
  • Cas Number : 1955505-50-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-184480

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-184480
Availability details for
Quantity
  • Availability
  • Add to Cart