ENA428682617 Enamine

6-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1,2,3,4-tetrahydroquinolin-2-one

  • MDL number MFCD18072003
  • Cas Number : 400620-72-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-119221

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-119221
Availability details for
Quantity
  • Availability
  • Add to Cart