ENA428682637 Enamine

5-(chloromethyl)-1-(2,2,2-trifluoroethyl)-1H-1,2,3,4-tetrazole

  • MDL number MFCD16863247
  • Cas Number : 1341047-82-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-179411

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-179411
Availability details for
Quantity
  • Availability
  • Add to Cart