ENA428682638 Enamine

5-(chloromethyl)-1-[2-(2,2,2-trifluoroethoxy)ethyl]-1H-1,2,3,4-tetrazole

  • MDL number MFCD18894665
  • Cas Number : 1342882-56-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-179412

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-179412
Availability details for
Quantity
  • Availability
  • Add to Cart