ENA428682639 Enamine

3-[4-(trifluoromethyl)phenyl]-1,2,4-thiadiazol-5-amine

  • MDL number MFCD12137274
  • Cas Number : 1153981-93-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-179493

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-179493
Availability details for
Quantity
  • Availability
  • Add to Cart