ENA428682640 Enamine

[3-(pyridin-2-yloxy)phenyl]methanol

  • MDL number MFCD09025855
  • Cas Number : 869901-22-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-179537

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-179537
Availability details for
Quantity
  • Availability
  • Add to Cart