ENA428682642 Enamine

1-ethyl-2-(1-methyl-1H-pyrazol-4-yl)-6-oxopiperidine-3-carboxylic acid

  • MDL number MFCD19416389
  • Cas Number : 1496200-27-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-179541

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-179541
Availability details for
Quantity
  • Availability
  • Add to Cart