ENA428682644 Enamine

3-(1,4-oxazepan-4-yl)propan-1-amine

  • MDL number MFCD17149551
  • Cas Number : 49809-08-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-179556

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-179556
Availability details for
Quantity
  • Availability
  • Add to Cart