ENA428682652 Enamine

N,N-dimethylazepan-4-amine

  • MDL number MFCD18917058
  • Cas Number : 933704-58-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-179770

Safety Information

signalword Danger
pcodes P210,P233,P240,P241,P242,P243,P261,P264,P270,P271,P280,P301+P312,P302+P352,P303+P361+P353,P304+P340,P305+P351+P338,P310,P312,P330,P332+P313,P362,P370+P378,P403+P233,P403+P235,P405,P501
hcodes Xn,F
MSDS https://www.enaminestore.com/catalog/EN300-179770
pictograms GHS02,GHS05,GHS07
hstatements H225,H302,H315,H318,H335
Availability details for
Quantity
  • Availability
  • Add to Cart