ENA428682657 Enamine

2-(propan-2-yl)-3,4-dihydroquinazoline-4-thione

  • MDL number MFCD03713577
  • Cas Number : 13193-53-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-179813

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-179813
Availability details for
Quantity
  • Availability
  • Add to Cart