ENA428682661 Enamine

3-chloro-6-(propan-2-yl)pyridazine

  • MDL number MFCD12143153
  • Cas Number : 570416-35-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 32-33
Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-179890

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-179890
Availability details for
Quantity
  • Availability
  • Add to Cart