ENA428682685 Enamine

6-chloro-2-cyclopropyl-1,4-dihydroquinolin-4-one

  • MDL number MFCD18338008
  • Cas Number : 1341362-65-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-180120

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-180120
Availability details for
Quantity
  • Availability
  • Add to Cart