ENA428682689 Enamine

2,4-dipropoxybenzoic acid

  • MDL number MFCD12415011
  • Cas Number : 695812-18-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-180161

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-180161
Availability details for
Quantity
  • Availability
  • Add to Cart