ENA428682692 Enamine

[(2-bromo-5-methoxyphenyl)methyl](methyl)amine

  • MDL number MFCD11105972
  • Cas Number : 767289-08-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-180185

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-180185
Availability details for
Quantity
  • Availability
  • Add to Cart