ENA428682693 Enamine

[(2,5-dimethylphenyl)methyl](methyl)amine

  • MDL number MFCD11204737
  • Cas Number : 1079178-90-7
  • Molecular Weight : 149.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-180186

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-180186
Availability details for
Quantity
  • Availability
  • Add to Cart