ENA428682707 Enamine

6-(cyclopropylcarbamoyl)cyclohex-3-ene-1-carboxylic acid

  • MDL number MFCD03361803
  • Cas Number : 540521-54-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-180296

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-180296
Availability details for
Quantity
  • Availability
  • Add to Cart