ENA428682717 Enamine

4-(but-3-yn-1-yl)morpholine

  • MDL number MFCD18250546
  • Cas Number : 14731-39-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-180347
pictograms GHS05,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart