ENA428682720 Enamine

1-(3-cyanopropyl)-1H-indole-3-carboxylic acid

  • MDL number MFCD11155410
  • Cas Number : 1020984-72-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-180399

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-180399
Availability details for
Quantity
  • Availability
  • Add to Cart