ENA428682722 Enamine

2-(2-fluorophenyl)-1-phenylethan-1-one

  • MDL number MFCD02260682
  • Cas Number : 3826-47-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-180402

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-180402
Availability details for
Quantity
  • Availability
  • Add to Cart