ENA428682725 Enamine

1-(pyridazin-3-yl)piperidin-4-amine

  • MDL number MFCD16767464
  • Cas Number : 898271-24-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-180423

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-180423
Availability details for
Quantity
  • Availability
  • Add to Cart