ENA428682729 Enamine

6-ethyl-2-(4-methylphenyl)-3,4-dihydropyrimidin-4-one

  • MDL number MFCD12137537
  • Cas Number : 1154549-27-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-180427

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-180427
Availability details for
Quantity
  • Availability
  • Add to Cart