ENA428682748 Enamine

2,2-dimethylheptanedinitrile

  • MDL number MFCD28118196
  • Cas Number : 500310-59-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-136623

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-136623
Availability details for
Quantity
  • Availability
  • Add to Cart