ENA428682749 Enamine

2,6-dibromo-4-(2-hydroxyethyl)phenol

  • MDL number MFCD27939847
  • Cas Number : 303009-05-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-180521

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-180521
Availability details for
Quantity
  • Availability
  • Add to Cart