ENA428682755 Enamine

5-(4-nitrophenyl)-1,2,4-oxadiazol-3-amine

  • MDL number MFCD18867056
  • Cas Number : 23275-50-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-180591

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-180591
Availability details for
Quantity
  • Availability
  • Add to Cart