ENA428682756 Enamine

4-(5-methyl-1,3-thiazol-2-yl)piperidine

  • MDL number MFCD16834289
  • Cas Number : 1249151-01-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-180592

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-180592
Availability details for
Quantity
  • Availability
  • Add to Cart