ENA428682760 Enamine

1-methyl-N-(propan-2-yl)-1H-imidazol-2-amine

  • MDL number MFCD08704752
  • Cas Number : 939758-36-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-180612

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-180612
Availability details for
Quantity
  • Availability
  • Add to Cart