ENA428682768 Enamine

5-(chloromethyl)-1-(oxan-4-yl)-1H-1,2,3,4-tetrazole

  • MDL number MFCD16863276
  • Cas Number : 1340126-75-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-180651

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-180651
Availability details for
Quantity
  • Availability
  • Add to Cart