ENA428682777 Enamine

5-{[4-(cyclopropylmethyl)piperazin-1-yl]methyl}pyridin-2-amine

  • MDL number MFCD17039070
  • Cas Number : 1457516-30-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-180746

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-180746
Availability details for
Quantity
  • Availability
  • Add to Cart