ENA428682780 Enamine

3-bromo-1-(3-bromophenyl)pyrrolidin-2-one

  • MDL number MFCD18886478
  • Cas Number : 1342077-83-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-180780

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-180780
Availability details for
Quantity
  • Availability
  • Add to Cart