ENA428682783 Enamine

2-(4-chlorophenyl)-5-(propan-2-yl)-1H-imidazole-4-carboxylic acid

  • MDL number MFCD12069994
  • Cas Number : 1156076-19-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-180844

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-180844
Availability details for
Quantity
  • Availability
  • Add to Cart